ENAH5802E7D5 Enamine

imidazo[2,1-b][1,3]thiazole-2-carboxylic acid hydrochloride

  • Cas Number : 409062-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443438
Availability details for
Quantity
  • Availability
  • Add to Cart