ENAH5802E7D8 Enamine

(2Z)-3,7-dimethylocta-2,6-dienoic acid

  • Cas Number : 4613-38-1
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7429959

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7429959
pictograms GHS07
hstatements H312,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart