ENAH5802E7DA Enamine

2-carboxy-4-methoxypyridin-1-ium-1-olate

  • MDL number MFCD14581994
  • Cas Number : 17209-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426196
Availability details for
Quantity
  • Availability
  • Add to Cart