ENAH5802E7DC Enamine

1-cyclopropyl-5-oxopyrrolidine-2-carboxylic acid

  • MDL number MFCD15730151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3044054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3044054
Availability details for
Quantity
  • Availability
  • Add to Cart