ENAH5802E7DF Enamine

lithium(1+) 6-oxo-5-oxa-7-azaspiro[3.4]octane-8-carboxylate

  • MDL number MFCD32066542
  • Cas Number : 2361635-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432415
Availability details for
Quantity
  • Availability
  • Add to Cart