ENAH5802E7E2 Enamine

1-methylcyclooctane-1-carboxylic acid

  • MDL number MFCD20643679
  • Cas Number : 35664-94-9
  • Molecular Weight : 170.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 45-47
Iupac Name 1-methylcyclooctanecarboxylic acid
Inchi Code 1S/C10H18O2/c1-10(9(11)12)7-5-3-2-4-6-8-10/h2-8H2,1H3,(H,11,12)
Inchi Key NVHVUKCFHWOQQR-UHFFFAOYSA-N

Description

Melting Point 45-47
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-3025921

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-3025921
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart