ENAH5802E7E3 Enamine

sodium 3,3,3-trifluoro-2-methoxy-2-methylpropanoate

  • Cas Number : 2344680-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425241
Availability details for
Quantity
  • Availability
  • Add to Cart