ENAH5802E7E5 Enamine

3-hydroxy-1-azabicyclo[2.2.2]octane-3-carboxylic acid hydrochloride

  • Cas Number : 98426-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442191
Availability details for
Quantity
  • Availability
  • Add to Cart