ENAH5802E7E9 Enamine

3-(dimethylamino)cyclohexane-1-carboxylic acid hydrochloride

  • MDL number MFCD32198579
  • Cas Number : 1580507-71-6
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(dimethylamino)cyclohexane-1-carboxylic acid hydrochloride
Inchi Code 1S/C9H17NO2.ClH/c1-10(2)8-5-3-4-7(6-8)9(11)12;/h7-8H,3-6H2,1-2H3,(H,11,12);1H
Inchi Key TUWBFBHPFUGYMV-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437176
Availability details for
Quantity
  • Availability
  • Add to Cart