ENAH5802E7EA Enamine

2-{hexahydrofuro[3,2-b]furan-2-yl}acetic acid

  • Cas Number : 2344678-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355519
Availability details for
Quantity
  • Availability
  • Add to Cart