ENAH5802E7EE Enamine

2-ethyl-2,5-dimethylhexanoic acid

  • MDL number MFCD16855768
  • Cas Number : 24353-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430024
Availability details for
Quantity
  • Availability
  • Add to Cart