ENAH5802E7F0 Enamine

3-(difluoromethyl)pyridine-4-carboxylic acid

  • MDL number MFCD18257234
  • Cas Number : 1211541-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356271
Availability details for
Quantity
  • Availability
  • Add to Cart