ENAH5802E7F6 Enamine

2-(4-methoxyoxan-4-yl)acetic acid

  • MDL number MFCD30200938
  • Cas Number : 1785066-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177458
Availability details for
Quantity
  • Availability
  • Add to Cart