ENAH5802E7F8 Enamine

(2S)-2-(carbamoylamino)-4-methylpentanoic acid

  • MDL number MFCD00057656
  • Cas Number : 26117-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-205
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4207624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4207624
Availability details for
Quantity
  • Availability
  • Add to Cart