ENAH5802E7FA Enamine

lithium(1+) 7-methyl-[1,2,4]triazolo[4,3-a]pyridine-3-carboxylate

  • MDL number MFCD32066227
  • Cas Number : 2361645-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434072
Availability details for
Quantity
  • Availability
  • Add to Cart