ENAH5802E7FF Enamine

3-methylimidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride

  • Cas Number : 2375271-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440211
Availability details for
Quantity
  • Availability
  • Add to Cart