ENAH5802E801 Enamine

lithium(1+) ion 2,2-dioxo-2lambda6-thia-1-azabicyclo[2.1.1]hexane-4-carboxylate

  • MDL number MFCD31924413
  • Cas Number : 2344679-85-0
  • Molecular Weight : 183.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name lithium 2-thia-1-azabicyclo[2.1.1]hexane-4-carboxylate 2,2-dioxide
Inchi Code 1S/C5H7NO4S.Li/c7-4(8)5-1-6(2-5)11(9,10)3-5;/h1-3H2,(H,7,8);/q;+1/p-1
Inchi Key YWNUEYFXJBMLKV-UHFFFAOYSA-M

Description

Purity 95
Salt Data [Li+]
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7425987

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7425987
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart