ENAH5802E803 Enamine

3-methylpyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD24503590
  • Cas Number : 1398584-50-3
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methylpyrazolo[1,5-a]pyridine-2-carboxylic acid
Inchi Code 1S/C9H8N2O2/c1-6-7-4-2-3-5-11(7)10-8(6)9(12)13/h2-5H,1H3,(H,12,13)
Inchi Key GELYDKXMSHXTOJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432559

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart