ENAH5802E805 Enamine

8-methylimidazo[1,2-a]pyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD28989846
  • Cas Number : 2344685-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763388
Availability details for
Quantity
  • Availability
  • Add to Cart