ENAH5802E806 Enamine

3,3-dioxo-3lambda6-thiabicyclo[3.1.0]hexane-6-carboxylic acid

  • MDL number MFCD26938531
  • Cas Number : 1783317-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6779469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6779469
Availability details for
Quantity
  • Availability
  • Add to Cart