ENAH5802E809 Enamine

1-methyl-2,3-dihydro-1H-indene-2-carboxylic acid

  • MDL number MFCD20639454
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-2-indanecarboxylic acid
Inchi Code 1S/C11H12O2/c1-7-9-5-3-2-4-8(9)6-10(7)11(12)13/h2-5,7,10H,6H2,1H3,(H,12,13)
Inchi Key IHSLASJHLBMFJC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100609
Availability details for
Quantity
  • Availability
  • Add to Cart