ENAH5802E80C Enamine

3-methyl-1H-pyrazolo[4,3-c]pyridine-6-carboxylic acid hydrochloride

  • MDL number MFCD32069181
  • Cas Number : 2375259-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438571
Availability details for
Quantity
  • Availability
  • Add to Cart