ENAH5802E80F Enamine

(2Z)-3-chloro-5-(dimethylamino)pent-2-enoic acid hydrochloride

  • MDL number MFCD32068668
  • Cas Number : 2361914-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434623
Availability details for
Quantity
  • Availability
  • Add to Cart