ENAH5802E814 Enamine

5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD26939071
  • Cas Number : 1691859-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21221
Availability details for
Quantity
  • Availability
  • Add to Cart