ENAH5802E816 Enamine

5,6,7,8-tetrahydrocinnoline-3-carboxylic acid hydrochloride

  • MDL number MFCD32068843
  • Cas Number : 2361723-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7436026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436026
Availability details for
Quantity
  • Availability
  • Add to Cart