ENAH5802E817 Enamine

2-hydroxy-1H-imidazo[4,5-c]pyridine-7-carboxylic acid

  • MDL number MFCD32068897
  • Cas Number : 1782640-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434209
Availability details for
Quantity
  • Availability
  • Add to Cart