ENAH5802E818 Enamine

5-(1,3-oxazol-2-yl)furan-2-carboxylic acid

  • MDL number MFCD20644851
  • Cas Number : 106833-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229-231
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2148883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2148883
Availability details for
Quantity
  • Availability
  • Add to Cart