ENAH5802E819 Enamine

5-hydroxyimidazo[1,5-c]pyrimidine-7-carboxylic acid

  • MDL number MFCD28520374
  • Cas Number : 1784884-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443518
Availability details for
Quantity
  • Availability
  • Add to Cart