ENAH5802E81B Enamine

3-(furan-2-yl)-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD07701227
  • Cas Number : 901927-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-295365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-295365
Availability details for
Quantity
  • Availability
  • Add to Cart