ENAH5802E81D Enamine

5-carbamoyl-2-methylbenzoic acid

  • MDL number MFCD20652200
  • Cas Number : 99357-88-7
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 255-258
Iupac Name 5-(aminocarbonyl)-2-methylbenzoic acid
Inchi Code 1S/C9H9NO3/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4H,1H3,(H2,10,11)(H,12,13)
Inchi Key LZMONZCDSLCIAC-UHFFFAOYSA-N

Description

Melting Point 255-258
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-379290

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-379290
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart