ENAH5802E81F Enamine

lithium(1+) 2-methyl-5H,6H,7H,8H-imidazo[1,2-a]pyridine-3-carboxylate

  • Cas Number : 2375274-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439763
Availability details for
Quantity
  • Availability
  • Add to Cart