ENAH5802E825 Enamine

[1,3]thiazolo[5,4-b]pyridine-6-carboxylic acid

  • MDL number MFCD18256939
  • Cas Number : 1256804-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1601940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1601940
Availability details for
Quantity
  • Availability
  • Add to Cart