ENAH5802E826 Enamine

3-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD32069229
  • Cas Number : 1782861-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438912
Availability details for
Quantity
  • Availability
  • Add to Cart