ENAH5802E827 Enamine

2-ethoxy-5-methylbenzoic acid

  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-ethoxy-5-methylbenzoic acid
Inchi Code 1S/C10H12O3/c1-3-13-9-5-4-7(2)6-8(9)10(11)12/h4-6H,3H2,1-2H3,(H,11,12)
Inchi Key LRLFEBJGOYSOFO-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7442721
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart