ENAH5802E828 Enamine

2-methyl-4,5,6,7-tetrahydro-2H-indazole-6-carboxylic acid

  • Cas Number : 1546150-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158726
Availability details for
Quantity
  • Availability
  • Add to Cart