ENAH5802E82B Enamine

3,7-dimethyl-5H,6H,7H-pyrrolo[2,1-c][1,2,4]triazole-7-carboxylic acid hydrochloride

  • MDL number MFCD32066390
  • Cas Number : 2361635-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431629
Availability details for
Quantity
  • Availability
  • Add to Cart