ENAH5802E82D Enamine

3-methyl-4H-thieno[3,2-b]pyrrole-5-carboxylic acid

  • MDL number MFCD19688278
  • Cas Number : 332099-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3019086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3019086
Availability details for
Quantity
  • Availability
  • Add to Cart