ENAH5802E830 Enamine

1-methyl-1H-thieno[2,3-d]imidazole-5-carboxylic acid

  • MDL number MFCD22064476
  • Cas Number : 1369371-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437173
Availability details for
Quantity
  • Availability
  • Add to Cart