ENAH5802E832 Enamine

4,6-dimethoxypyridine-3-carboxylic acid

  • MDL number MFCD18255516
  • Cas Number : 1256807-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109983
Availability details for
Quantity
  • Availability
  • Add to Cart