ENAH5802E833 Enamine

tert-butyl 8-(aminomethyl)-5-azaspiro[3.5]nonane-5-carboxylate

  • Cas Number : 2375270-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426977
Availability details for
Quantity
  • Availability
  • Add to Cart