ENAH5802E834 Enamine

6-methylidene-5H,6H-[1,2,4]triazolo[3,4-b][1,3]thiazole-3-carboxylic acid

  • MDL number MFCD32066384
  • Cas Number : 2361635-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431518
Availability details for
Quantity
  • Availability
  • Add to Cart