ENAH5802E835 Enamine

1-(prop-2-enoyl)piperidine-4-carboxylic acid

  • MDL number MFCD09931332
  • Cas Number : 109486-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-878668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-878668
Availability details for
Quantity
  • Availability
  • Add to Cart