ENAH5802E838 Enamine

6-hydroxy-5H,6H,7H-pyrazolo[3,2-b][1,3]oxazine-3-carboxylic acid

  • MDL number MFCD29064519
  • Cas Number : 2091716-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6497444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6497444
Availability details for
Quantity
  • Availability
  • Add to Cart