ENAH5802E83B Enamine

5-(3-hydroxyoxetan-3-yl)-1,2-oxazole-3-carboxylic acid

  • Cas Number : 2092185-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-365017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365017
Availability details for
Quantity
  • Availability
  • Add to Cart