ENAH5802E83E Enamine

2-hydroxy-3-methyl-2-(trifluoromethyl)butanoic acid

  • MDL number MFCD28040214
  • Cas Number : 1823842-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425890
Availability details for
Quantity
  • Availability
  • Add to Cart