ENAH5802E83F Enamine

3-cyano-1H-indole-4-carboxylic acid

  • MDL number MFCD07776626
  • Cas Number : 889942-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7392579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7392579
Availability details for
Quantity
  • Availability
  • Add to Cart