ENAH5802E841 Enamine

1-hydroxy-4-propylcyclohexane-1-carboxylic acid

  • MDL number MFCD14602045
  • Cas Number : 1250909-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111306
Availability details for
Quantity
  • Availability
  • Add to Cart