ENAH5802E845 Enamine

3-cyano-1H-indazole-5-carboxylic acid

  • MDL number MFCD20646239
  • Cas Number : 1301214-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-716135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-716135
Availability details for
Quantity
  • Availability
  • Add to Cart