ENAH5802E846 Enamine

2-methyl-3-(morpholin-4-yl)-3-oxopropanoic acid

  • MDL number MFCD26600165
  • Cas Number : 1525711-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2855057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2855057
Availability details for
Quantity
  • Availability
  • Add to Cart