ENAH5802E84B Enamine

3-[(2,3-dimethylbutyl)(methyl)amino]propanoic acid hydrochloride

  • Cas Number : 2375273-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444255
Availability details for
Quantity
  • Availability
  • Add to Cart